Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008629.1 | NP_032655.1 | 9 - 10 | 710 | 68 | |
GenBank mRNA | AK084019.1 | - | 9 - 10 | 693 | 68 | |
D49654.1 | - | 9 - 10 | 710 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |