Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spatc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028852.1 | NP_083128.1 | 2 - 3 | 941 | 65 | |
GenBank mRNA | AB032199.1 | - | 2 - 3 | 941 | 65 | |
AK006994.1 | - | 2 - 3 | 942 | 65 | ||
BC118509.1 | - | 2 - 3 | 901 | 40106461 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |