Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026555.2 | NP_080831.2 | 1 - 2 | 116 | 62 | |
GenBank mRNA | AK077943.1 | - | 1 - 2 | 56 | 62 | |
AK086705.1 | - | 1 - 2 | 56 | 62 | ||
BC025602.1 | - | 1 - 2 | 67 | 5250505 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |