Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026555.2 | NP_080831.2 | 3 - 4 | 571 | 62 | |
GenBank mRNA | AK003918.1 | - | 2 - 3 | 241 | 62 | |
AK077943.1 | - | 3 - 4 | 511 | 62 | ||
AK082762.1 | - | 3 - 4 | 501 | 62 | ||
AK086705.1 | - | 3 - 4 | 511 | 62 | ||
AK132143.1 | - | 3 - 4 | 503 | 62 | ||
BC005487.1 | - | 2 - 3 | 408 | 62 | ||
BC025602.1 | - | 3 - 4 | 522 | 5250505 | 62 | |
BC055903.1 | - | 3 - 4 | 518 | 4010685 | 62 | |
BC092069.1 | - | 3 - 4 | 469 | 4215821 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |