Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030233.1 | NP_084509.1 | 9 - 10 | 1062 | 109 | |
GenBank mRNA | AB489137.1 | - | 9 - 10 | 1043 | 109 | |
AK019878.1 | - | 9 - 10 | 1063 | 109 | ||
AK030833.1 | - | 9 - 10 | 1068 | 109 | ||
AK031128.1 | - | 7 - 8 | 2910 | 109 | ||
AK033202.1 | - | 9 - 10 | 1073 | 109 | ||
AK035874.1 | - | 8 - 9 | 3106 | 109 | ||
AK132193.1 | - | 9 - 10 | 1063 | 109 | ||
AK134418.1 | - | 9 - 10 | 1070 | 109 | ||
BC019985.1 | - | 10 - 11 | 1412 | 109 | ||
BC028473.1 | - | 4 - 5 | 476 | 109 | ||
BC068127.1 | - | 9 - 10 | 1059 | 109 | ||
BC087944.1 | - | 9 - 10 | 1074 | 30314036 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |