Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700026794.mm.1 |
NM_175022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026794.mm.1 |
NM_175022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026794.mm.1 |
TC0700002984.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026795.mm.1 |
NM_175022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026795.mm.1 |
NM_175022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026795.mm.1 |
TC0700002984.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026796.mm.1 |
NM_175022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026796.mm.1 |
NM_175022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026796.mm.1 |
TC0700002984.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026797.mm.1 |
NM_175022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026797.mm.1 |
NM_175022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026797.mm.1 |
TC0700002984.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700026793.mm.1 |
NM_175022.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026793.mm.1 |
NM_175022.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700026793.mm.1 |
TC0700002984.mm.1 |
MTA TC |