Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013468.3 | NP_038496.2 | 2 - 3 | 265 | 154 | |
GenBank mRNA | AF041847.1 | - | 2 - 3 | 216 | 154 | |
AK009655.1 | - | 2 - 3 | 258 | 154 | ||
AK009959.1 | - | 2 - 3 | 264 | 154 | ||
AK145944.1 | - | 2 - 3 | 306 | 154 | ||
AK146471.1 | - | 2 - 3 | 262 | 154 | ||
AK146627.1 | - | 2 - 3 | 264 | 154 | ||
AK146940.1 | - | 2 - 3 | 262 | 154 | ||
BC037138.1 | - | 2 - 3 | 265 | 4953125 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |