Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013468.3 | NP_038496.2 | 7 - 8 | 819 | 64 | |
GenBank mRNA | AF041847.1 | - | 7 - 8 | 770 | 64 | |
AK009655.1 | - | 7 - 8 | 812 | 64 | ||
AK009959.1 | - | 7 - 8 | 818 | 64 | ||
AK145944.1 | - | 7 - 8 | 860 | 64 | ||
AK146471.1 | - | 7 - 8 | 816 | 64 | ||
AK146627.1 | - | 7 - 8 | 818 | 64 | ||
AK146940.1 | - | 7 - 8 | 816 | 64 | ||
BC037138.1 | - | 7 - 8 | 819 | 4953125 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |