Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010615.1 | NP_034745.1 | 11 - 12 | 1486 | 135 | |
GenBank mRNA | AJ223293.1 | - | 10 - 11 | 1181 | 135 | |
AK083324.1 | - | 11 - 12 | 1463 | 135 | ||
AK147754.1 | - | 11 - 12 | 1510 | 135 | ||
BC060670.1 | - | 11 - 12 | 1486 | 6406754 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |