Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198163.1 | NP_937806.1 | 4 - 5 | 514 | 63 | |
GenBank mRNA | AB232631.1 | - | 4 - 5 | 354 | 63 | |
AK156576.1 | - | 4 - 5 | 505 | 63 | ||
BC030941.1 | - | 2 - 3 | 205 | 63 | ||
BC056466.1 | - | 4 - 5 | 514 | 5717246 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |