Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008846.2 | NP_032872.1 | 9 - 10 | 1214 | 80 | |
GenBank mRNA | AK133809.1 | - | 9 - 10 | 1209 | 80 | |
AK146100.1 | - | 9 - 10 | 1214 | 80 | ||
AK149354.1 | - | 9 - 10 | 1214 | 80 | ||
AK164552.1 | - | 9 - 10 | 1197 | 80 | ||
BC034864.1 | - | 9 - 10 | 1228 | 3326897 | 80 | |
D86176.1 | - | 8 - 9 | 1147 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |