Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026805.1 | NP_081081.1 | 2 - 3 | 391 | 57 | |
GenBank mRNA | AK047655.1 | - | 2 - 3 | 382 | 57 | |
AK047764.1 | - | 2 - 3 | 392 | 57 | ||
AK053833.1 | - | 2 - 3 | 382 | 57 | ||
AK082744.1 | - | 2 - 3 | 393 | 57 | ||
AK160770.1 | - | 2 - 3 | 382 | 57 | ||
BC059878.1 | - | 2 - 3 | 344 | 6812082 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |