Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198109.4 | NP_932777.2 | 1 - 2 | 98 | 66 | |
GenBank mRNA | AB099287.1 | - | 1 - 2 | 133 | 66 | |
AK089308.1 | - | 1 - 2 | 102 | 66 | ||
AK155557.1 | - | 1 - 2 | 123 | 66 | ||
AK170787.1 | - | 1 - 2 | 88 | 66 | ||
AK171210.1 | - | 1 - 2 | 147 | 66 | ||
AK171556.1 | - | 1 - 2 | 113 | 66 | ||
AK173144.1 | - | 1 - 2 | 120 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |