Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: As3mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020577.2 | NP_065602.2 | 2 - 3 | 132 | 73 | |
GenBank mRNA | AF166383.1 | - | 2 - 3 | 102 | 73 | |
AK006571.1 | - | 2 - 3 | 117 | 73 | ||
AK009814.1 | - | 1 - 2 | 119 | 73 | ||
AK011320.1 | - | 2 - 3 | 84 | 73 | ||
BC013468.1 | - | 2 - 3 | 57 | 73 | ||
BC145663.1 | - | 2 - 3 | 74 | 40130966 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |