Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027156.3 | NP_081432.2 | 3 - 4 | 618 | 63 | |
GenBank mRNA | AK141361.1 | - | 3 - 4 | 598 | 63 | |
AK152599.1 | - | 3 - 4 | 586 | 63 | ||
BC060646.1 | - | 3 - 4 | 595 | 6853826 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |