Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077264.1 | NP_001070732.1 | 10 - 11 | 1418 | 74 | |
NM_007458.2 | NP_031484.1 | 10 - 11 | 1418 | 74 | ||
GenBank mRNA | AK135424.1 | - | 10 - 11 | 1347 | 74 | |
AK155515.1 | - | 10 - 11 | 1355 | 74 | ||
AK200363.1 | - | 1 - 2 | 80 | 74 | ||
BC031433.1 | - | 10 - 11 | 1368 | 4481643 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |