Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013909.2 | NP_038937.2 | 4 - 5 | 779 | 66 | |
GenBank mRNA | AB041571.1 | - | 3 - 4 | 1017 | 66 | |
AF176522.1 | - | 4 - 5 | 767 | 66 | ||
AK040458.1 | - | 2 - 3 | 1159 | 66 | ||
BC011485.1 | - | 2 - 3 | 1152 | 66 | ||
BC026818.1 | - | 2 - 3 | 1146 | 66 | ||
BC032263.1 | - | 5 - 6 | 66 | |||
BC044784.2 | - | 3 - 4 | 979 | 66 | ||
BC054377.1 | - | 3 - 4 | 600 | 6315438 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |