Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500004535.mm.1 |
NM_001163621.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004535.mm.1 |
NM_001163621.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004535.mm.1 |
NM_028010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004535.mm.1 |
NM_028010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004535.mm.1 |
TC1500000617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004532.mm.1 |
NM_028010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004532.mm.1 |
NM_028010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004532.mm.1 |
TC1500000617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004533.mm.1 |
NM_028010.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004533.mm.1 |
NM_028010.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004533.mm.1 |
TC1500000617.mm.1 |
MTA TC |