Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Recql4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058214.3 | NP_478121.2 | 2 - 3 | 205 | 57 | |
GenBank mRNA | AB039882.1 | - | 2 - 3 | 78 | 57 | |
AB175741.1 | - | 2 - 3 | 122 | 57 | ||
AK090260.1 | - | 2 - 3 | 195 | 57 | ||
BC064680.1 | - | 2 - 3 | 179 | 57 | ||
BC131656.1 | - | 2 - 3 | 118 | 57 | ||
BC131657.1 | - | 2 - 3 | 113 | 57 | ||
BY122921.1 | - | 2 - 3 | 205 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |