Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019982.3 | NP_064366.2 | 13 - 14 | 2945 | 59 | |
GenBank mRNA | AK083229.1 | - | 13 - 14 | 2909 | 59 | |
BC065117.1 | - | 13 - 14 | 2905 | 6840338 | 59 | |
DQ167195.1 | - | 13 - 14 | 2932 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |