Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2w1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160265.1 | NP_001153737.1 | 5 - 6 | 855 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |