Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aknad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177859.3 | NP_808527.2 | 5 - 6 | 1543 | 104 | |
GenBank mRNA | AK135678.1 | - | 4 - 5 | 503 | 104 | |
AK164420.1 | - | 5 - 6 | 1546 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |