Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053158.3 | NP_444388.2 | 7 - 8 | 842 | 65 | |
GenBank mRNA | AB049632.1 | - | 6 - 7 | 798 | 65 | |
AK010343.1 | - | 7 - 8 | 816 | 65 | ||
AK083978.1 | - | 7 - 8 | 818 | 65 | ||
AK142249.1 | - | 7 - 8 | 817 | 65 | ||
BC028774.1 | - | 1 - 2 | 88 | 65 | ||
BC061042.1 | - | 7 - 8 | 806 | 6706599 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |