Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF128212.1 | - | 4 - 5 | 335 | 77 | |
AF128213.1 | - | 4 - 5 | 335 | 77 | ||
BC116851.1 | - | 4 - 5 | 339 | 77 | ||
BC116855.1 | - | 4 - 5 | 339 | 77 | ||
M86672.1 | - | 4 - 5 | 437 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |