Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133653.3 | NP_598414.1 | 6 - 7 | 1024 | 74 | |
GenBank mRNA | AK149542.1 | - | 5 - 6 | 775 | 74 | |
AK153832.1 | - | 5 - 6 | 778 | 74 | ||
BC011211.1 | - | 5 - 6 | 762 | 4196928 | 74 | |
L13622.1 | - | 5 - 6 | 776 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |