Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172711.2 | NP_766299.1 | 13 - 14 | 1665 | 100 | |
GenBank mRNA | AK084627.1 | - | 13 - 14 | 1646 | 100 | |
BC039660.1 | - | 6 - 7 | 749 | 100 | ||
BC057962.1 | - | 11 - 12 | 1355 | 5353891 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |