Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wapal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004436.4 | NP_001004436.2 | 8 - 9 | 2619 | 112 | |
NM_001301330.1 | NP_001288259.1 | 8 - 9 | 2601 | 112 | ||
GenBank mRNA | AB167349.1 | - | 8 - 9 | 2425 | 112 | |
AK028167.1 | - | 6 - 7 | 723 | 112 | ||
AK129105.1 | - | 8 - 9 | 2619 | 112 | ||
BC037674.1 | - | 2 - 3 | 166 | 112 | ||
BC139434.1 | - | 8 - 9 | 2374 | 8862456 | 112 | |
BC145625.1 | - | 8 - 9 | 2356 | 9054193 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |