Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039071.2 | NP_001034160.1 | 3 - 4 | 358 | 60 | |
NM_001039072.2 | NP_001034161.1 | 3 - 4 | 358 | 60 | ||
NM_001039073.2 | NP_001034162.1 | 3 - 4 | 358 | 60 | ||
NM_001039074.2 | NP_001034163.1 | 2 - 3 | 389 | 60 | ||
NM_001039075.2 | NP_001034164.1 | 3 - 4 | 358 | 60 | ||
NM_001039076.2 | NP_001034165.1 | 3 - 4 | 358 | 60 | ||
NM_011918.4 | NP_036048.3 | 3 - 4 | 358 | 60 | ||
GenBank mRNA | AF114378.2 | - | 3 - 4 | 324 | 60 | |
AF114379.2 | - | 3 - 4 | 322 | 60 | ||
AF228057.1 | - | 2 - 3 | 251 | 60 | ||
AF228058.1 | - | 2 - 3 | 251 | 60 | ||
AJ005621.1 | - | 3 - 4 | 351 | 60 | ||
AK004020.1 | - | 3 - 4 | 366 | 60 | ||
AK084461.1 | - | 3 - 4 | 338 | 60 | ||
AK137181.1 | - | 3 - 4 | 341 | 60 | ||
AK142292.1 | - | 3 - 4 | 310 | 60 | ||
AK172980.1 | - | 2 - 3 | 389 | 60 | ||
AY206011.1 | - | 2 - 3 | 275 | 60 | ||
AY206012.1 | - | 3 - 4 | 324 | 60 | ||
AY206013.1 | - | 3 - 4 | 322 | 60 | ||
AY206015.1 | - | 2 - 3 | 274 | 60 | ||
BC099596.1 | - | 3 - 4 | 325 | 60 | ||
BC138793.1 | - | 2 - 3 | 292 | 8861815 | 60 | |
BC145420.1 | - | 2 - 3 | 292 | 9053978 | 60 | |
BY704269.1 | - | 3 - 4 | 365 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |