Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011258.2 | NP_035388.2 | 9 - 10 | 1206 | 97 | |
GenBank mRNA | AK146164.1 | - | 9 - 10 | 1201 | 97 | |
AK146314.1 | - | 9 - 10 | 1201 | 97 | ||
BC085173.1 | - | 9 - 10 | 1183 | 30549691 | 97 | |
BC085174.1 | - | 9 - 10 | 1175 | 97 | ||
U01222.1 | - | 9 - 10 | 1174 | 97 | ||
U07157.1 | - | 9 - 10 | 1172 | 97 | ||
U15037.1 | - | 9 - 10 | 1207 | 97 | ||
U36441.1 | - | 9 - 10 | 1183 | 97 | ||
X72711.1 | - | 9 - 10 | 1172 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |