Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016786.3 | NP_058066.2 | 5 - 6 | 623 | 85 | |
GenBank mRNA | AB011081.1 | - | 5 - 6 | 561 | 85 | |
AK006316.1 | - | 2 - 3 | 148 | 85 | ||
AK018366.1 | - | 5 - 6 | 596 | 84 | ||
AK031055.1 | - | 3 - 4 | 323 | 85 | ||
AK035371.1 | - | 5 - 6 | 520 | 85 | ||
AK036068.1 | - | 5 - 6 | 564 | 85 | ||
AK048194.1 | - | 5 - 6 | 614 | 85 | ||
AK139564.1 | - | 5 - 6 | 640 | 85 | ||
AK160728.1 | - | 5 - 6 | 583 | 85 | ||
AK162142.1 | - | 5 - 6 | 564 | 85 | ||
AK166647.1 | - | 5 - 6 | 532 | 85 | ||
BC002013.1 | - | 5 - 6 | 471 | 3496393 | 85 | |
BC085311.1 | - | 5 - 6 | 549 | 30148009 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |