Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900003862.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003862.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003862.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003862.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003862.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003864.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003864.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003864.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003864.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003864.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003865.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003865.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003865.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003865.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003865.mm.1 |
TC1900000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003863.mm.1 |
NM_001161420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003863.mm.1 |
NM_001161420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003863.mm.1 |
NM_013703.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003863.mm.1 |
NM_013703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003863.mm.1 |
TC1900000415.mm.1 |
MTA TC |