Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113517.1 | NP_001106989.1 | 14 - 15 | 1732 | 66 | |
NM_001113518.1 | NP_001106990.1 | 13 - 14 | 1521 | 66 | ||
NM_017402.4 | NP_059098.2 | 13 - 14 | 1521 | 66 | ||
GenBank mRNA | AF247654.1 | - | 13 - 14 | 1417 | 66 | |
AF247655.1 | - | 11 - 12 | 1138 | 66 | ||
AF343877.1 | - | 13 - 14 | 1401 | 66 | ||
AK052545.1 | - | 13 - 14 | 1492 | 66 | ||
AK129064.1 | - | 14 - 15 | 1668 | 66 | ||
AK139156.1 | - | 14 - 15 | 1661 | 66 | ||
AK164308.1 | - | 1 - 2 | 2660 | 66 | ||
AY220301.1 | - | 13 - 14 | 1486 | 66 | ||
BC044838.1 | - | 13 - 14 | 1506 | 3499604 | 66 | |
U96634.1 | - | 11 - 12 | 1211 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |