Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025729.4 | NP_080005.2 | 5 - 6 | 2016 | 76 | |
GenBank mRNA | AK220185.1 | - | 5 - 6 | 2016 | 76 | |
BC139024.1 | - | 4 - 5 | 1873 | 8862046 | 76 | |
BC139025.1 | - | 4 - 5 | 1873 | 8862047 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |