Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009867.2 | NP_033997.1 | 5 - 6 | 873 | 71 | |
GenBank mRNA | BC047991.1 | - | 5 - 6 | 873 | 71 | |
D14888.1 | - | 5 - 6 | 783 | 71 | ||
X69966.1 | - | 5 - 6 | 729 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |