Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145482.3 | NP_663457.2 | 5 - 6 | 68 | ||
GenBank mRNA | AY037804.1 | - | 5 - 6 | 621 | 68 | |
BC029101.1 | - | 5 - 6 | 518 | 68 | ||
BY327995.1 | - | 3 - 4 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |