Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113389.1 | NP_001106860.1 | 7 - 8 | 1087 | 78 | |
NM_007890.2 | NP_031916.1 | 7 - 8 | 1346 | 78 | ||
GenBank mRNA | AK047845.1 | - | 2 - 3 | 334 | 78 | |
AK052390.1 | - | 1 - 2 | 222 | 78 | ||
BC034550.1 | - | 2 - 3 | 432 | 78 | ||
BC129888.1 | - | 6 - 7 | 949 | 40111469 | 78 | |
BC129889.1 | - | 6 - 7 | 949 | 40111470 | 78 | |
BC138716.1 | - | 6 - 7 | 976 | 8861738 | 78 | |
BC145405.1 | - | 6 - 7 | 949 | 9053963 | 78 | |
U58497.1 | - | 6 - 7 | 976 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |