Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178617.4 | NP_848732.3 | 9 - 10 | 963 | 76 | |
GenBank mRNA | AK039240.1 | - | 9 - 10 | 779 | 76 | |
AK050307.1 | - | 10 - 11 | 860 | 76 | ||
AK081951.1 | - | 9 - 10 | 855 | 76 | ||
AY278200.2 | - | 9 - 10 | 946 | 76 | ||
BC067055.1 | - | 9 - 10 | 963 | 30545678 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |