Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033457.2 | NP_001028629.2 | 6 - 7 | 1906 | 62 | |
GenBank mRNA | AK148406.1 | - | 6 - 7 | 1910 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |