Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026893.3 | NP_081169.2 | 2 - 3 | 670 | 70 | |
GenBank mRNA | AK005099.1 | - | 2 - 3 | 616 | 70 | |
AK030173.1 | - | 2 - 3 | 622 | 70 | ||
AK031089.1 | - | 2 - 3 | 600 | 70 | ||
AK090028.1 | - | 2 - 3 | 619 | 70 | ||
AK143635.1 | - | 2 - 3 | 623 | 70 | ||
AK173289.1 | - | 2 - 3 | 530 | 70 | ||
BC023852.1 | - | 2 - 3 | 606 | 5346910 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |