Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175138.4 | NP_780347.2 | 10 - 11 | 1102 | 82 | |
GenBank mRNA | AK030213.1 | - | 10 - 11 | 1036 | 82 | |
AK047808.1 | - | 10 - 11 | 1103 | 82 | ||
BC138785.1 | - | 10 - 11 | 1047 | 8861807 | 82 | |
BC138787.1 | - | 10 - 11 | 1047 | 8861809 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |