Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hopx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159900.1 | NP_001153372.1 | 1 - 2 | 117 | 56 | |
GenBank mRNA | AF492704.1 | - | 1 - 2 | 105 | 56 | |
AK003784.1 | - | 1 - 2 | 106 | 56 | ||
AK008297.1 | - | 1 - 2 | 84 | 56 | ||
AK008439.1 | - | 1 - 2 | 84 | 56 | ||
BC024546.1 | - | 1 - 2 | 101 | 4989797 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |