Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025691.1 | NP_079967.1 | 8 - 9 | 848 | 103 | |
GenBank mRNA | AK133784.1 | - | 8 - 9 | 849 | 103 | |
AK164641.1 | - | 2 - 3 | 206 | 103 | ||
BC003434.1 | - | 8 - 9 | 836 | 103 | ||
BC019196.1 | - | 8 - 9 | 841 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |