Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190734.1 | NP_001177663.1 | 10 - 11 | 797 | 105 | |
NM_178896.5 | NP_849227.1 | 9 - 10 | 862 | 105 | ||
GenBank mRNA | AK033559.1 | - | 9 - 10 | 807 | 105 | |
AK076991.1 | - | 10 - 11 | 798 | 105 | ||
BC060213.1 | - | 9 - 10 | 804 | 105 | ||
BC108346.1 | - | 9 - 10 | 764 | 6310392 | 105 | |
BC137627.1 | - | 9 - 10 | 806 | 8860646 | 105 | |
BC137628.1 | - | 9 - 10 | 806 | 8860647 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |