Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159573.1 | NP_001153045.1 | 7 - 8 | 698 | 147 | |
NM_001159574.1 | NP_001153046.1 | 7 - 8 | 698 | 147 | ||
NM_024183.5 | NP_077145.2 | 7 - 8 | 698 | 147 | ||
GenBank mRNA | AK005061.1 | - | 7 - 8 | 667 | 147 | |
AK008561.1 | - | 7 - 8 | 670 | 147 | ||
AK049672.1 | - | 7 - 8 | 684 | 147 | ||
AK164105.1 | - | 7 - 8 | 664 | 147 | ||
BC003263.1 | - | 7 - 8 | 656 | 3499130 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |