Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008432.3 | NP_032458.3 | 7 - 8 | 767 | 132 | |
GenBank mRNA | AF039213.2 | - | 7 - 8 | 734 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mus musculus Probe spans exons |