Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163432.1 | NP_001156904.1 | 1 - 2 | 299 | 80 | |
NM_007712.3 | NP_031738.2 | 1 - 2 | 299 | 80 | ||
GenBank mRNA | AK148377.1 | - | 1 - 2 | 286 | 80 | |
BC015080.1 | - | 1 - 2 | 287 | 3995512 | 80 | |
BY021174.1 | - | 1 - 2 | 299 | 80 | ||
BY738871.1 | - | 1 - 2 | 259 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |