Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepsecs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172490.3 | NP_766078.1 | 8 - 9 | 1039 | 83 | |
GenBank mRNA | AK033654.1 | - | 8 - 9 | 1040 | 83 | |
AK049338.1 | - | 1046 | 83 | |||
AK153792.1 | - | 979 | 83 | |||
BC062133.1 | - | 8 - 9 | 1029 | 6516578 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |