Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042620.2 | NP_001036085.1 | 3 - 4 | 902 | 105 | |
NM_007839.3 | NP_031865.2 | 3 - 4 | 902 | 105 | ||
NR_104311.1 | 3 - 4 | 902 | 105 | |||
GenBank mRNA | AF017153.1 | - | 3 - 4 | 750 | 105 | |
AK145983.1 | - | 3 - 4 | 852 | 105 | ||
AK146441.1 | - | 3 - 4 | 848 | 105 | ||
AK149869.1 | - | 3 - 4 | 864 | 105 | ||
AK151015.1 | - | 3 - 4 | 865 | 105 | ||
AK151757.1 | - | 3 - 4 | 865 | 105 | ||
AK167927.1 | - | 3 - 4 | 775 | 105 | ||
AK168835.1 | - | 3 - 4 | 870 | 105 | ||
AK169174.1 | - | 3 - 4 | 870 | 105 | ||
BC003745.1 | - | 3 - 4 | 844 | 105 | ||
BC100343.1 | - | 3 - 4 | 845 | 30935044 | 105 | |
BC144759.1 | - | 3 - 4 | 733 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |