Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgfac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019447.2 | NP_062320.2 | 5 - 6 | 630 | 103 | |
GenBank mRNA | AF099017.1 | - | 5 - 6 | 590 | 103 | |
AF224724.1 | - | 5 - 6 | 590 | 103 | ||
AK007987.1 | - | 5 - 6 | 628 | 103 | ||
AK075718.1 | - | 5 - 6 | 539 | 103 | ||
AK144554.1 | - | 5 - 6 | 602 | 103 | ||
AK149522.1 | - | 5 - 6 | 594 | 103 | ||
BC019376.1 | - | 5 - 6 | 595 | 5134851 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |