Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008406.3 | NP_032432.2 | 15 - 16 | 1915 | 67 | |
GenBank mRNA | AK149500.1 | - | 15 - 16 | 1916 | 67 | |
BC013465.1 | - | 15 - 16 | 1899 | 67 | ||
BC028814.1 | - | 15 - 16 | 1915 | 5099284 | 67 | |
X70391.1 | - | 15 - 16 | 1895 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |